2,5-Dihydroxybenzoic acid, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,5-DHBA
, 2,5-Dihydroxybenzoic Acid
, Gentisic acid
, Hydroquinonecarboxylic acid
, DHB
M-Clarity™ quality level = MQ100
39319-10X10MG-EA
269
GBP
39319-10X10MG-
2,5-Dihydroxybenzoic acid, Supelco®
2,5-Dihydroxybenzoic acid
Formula:
C₇H₆O₄ MW: 154.12 g/mol Boiling Pt: 339 °C (1013 hPa) Melting Pt: 202…206 °C Density: 1.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002460 CAS Number: 490-79-9 EINECS: 207-718-5 Merck Index: 13,04409 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...