4,5-Dihydroxy-m-anisic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-O-Methylgallic acid, 3,4-Dihydroxy-5-methoxybenzoic acid, 3,4-Dihydroxy-5-Methoxy-Benzoic Acid, 3,4-Dihydroxy-5-methoxybenzenecarboxylic acid

013529-5G 013529-25G
FLUO013529-5GEA 0 GBP
FLUO013529-5G FLUO013529-25G
4,5-Dihydroxy-m-anisic acid
4,5-Dihydroxy-m-anisic acid
Formula: C₈H₈O₅
MW: 184.15 g/mol
MDL Number: MFCD00016518
CAS Number: 3934-84-7
EINECS: 223-512-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR