4-Nitrosalicylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Hydroxy-4-nitrobenzoic acid

M-Clarity™ quality level = MQ100

SIAL117188-25GEA 190 GBP
SIAL117188-25G
4-Nitrosalicylic acid, Sigma-Aldrich®
4-Nitrosalicylic acid
Formula: C₇H₅NO₅
MW: 183.12 g/mol
MDL Number: MFCD00071505
CAS Number: 619-19-2
EINECS: 210-584-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR