4-Aminobenzoic acid

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: PABAVitamin Bxpara-Aminobenzoic acidVitamin H1

SERA12400.01EA 0 GBP
SERA12400.01
4-Aminobenzoic acid
4-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR