2-Cyanobenzoic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MOLE38822788-1GEA 0 GBP
MOLE38822788-1G
2-Cyanobenzoic acid
2-Cyanobenzoic acid
Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00045796
CAS Number: 3839-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR