Dimethyl phthalate
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Phthalic acid dimethyl ester
ROTH4576.1EA
42.85
GBP
ROTH4576.1
Dimethyl phthalate
Dimethyl phthalate
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Boiling Pt: 283.7 °C (1013 hPa) Melting Pt: 6 °C Density: 1.192 g/cm³ (20 °C) Flash Pt: 146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008425 CAS Number: 131-11-3 EINECS: 205-011-6 Merck Index: 13,03281 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...