Methyl salicylate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Methyl 2-hydroxybenzoateWintergreen Oil

M-Clarity™ quality level = MQ100

SIAL84332-1LEA 110 GBP
SIAL84332-1L
Methyl salicylate, Sigma-Aldrich®
Methyl salicylate
Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 432 °C (1013 hPa)
Melting Pt: 100…101 °C
Density: 1.184 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 12,06200
https://uk.vwr.com/stibo/web/std.lang.all/89/31/2298931.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR