Isobutyl anthranilate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

I1315.25 I1315.100
XXXXI1315.25EA 0 GBP
XXXXI1315.25 XXXXI1315.100
Isobutyl anthranilate
Isobutyl anthranilate
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 156 °C (1013 hPa)
Density: 1,058 g/cm³ (20 °C)
MDL Number: MFCD00053630
CAS Number: 7779-77-3
EINECS: 231-938-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR