Ethyl-2-isothiocyanatobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-Ethoxycarbonylphenyl isothiocyanate, 2-Ethoxycarbonylphenyl isothiocyanate

E1474-E.10 E1474-E.50
XXXXE1474-E.10EA 0 GBP
XXXXE1474-E.10 XXXXE1474-E.50
Ethyl-2-isothiocyanatobenzoate
Ethyl 2-isothiocyanatobenzoate
Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Boiling Pt: 150…151 °C (1 mmHg)
Density: 1,201 g/cm³ (20 °C)
MDL Number: MFCD00041063
CAS Number: 99960-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR