3,4,5-Trimethoxybenzhydrazide

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4,5-Trimethoxybenzoic hydrazide, 3,4,5-Trimethoxybenzene-1-carbohydrazide, 3,4,5-Trimethoxybenzoic acid hydrazide, 3,4,5-Trimethoxybenzohydrazide

T2910.10 T2910.50
XXXXT2910.10EA 0 GBP
XXXXT2910.10 XXXXT2910.50
3,4,5-Trimethoxybenzhydrazide
3,4,5-Trimethoxybenzhydrazide
Formula: C₁₀H₁₄N₂O₄
MW: 226.23 g/mol
MDL Number: MFCD00014758
CAS Number: 3291-03-0
EINECS: 221-954-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR