3-Ethoxybenzoyl chloride ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H50227.MDEA
0
GBP
H50227.MD
H50227.03
H50227.14
3-Ethoxybenzoyl chloride ≥98%
3-Ethoxybenzoyl chloride
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00000677 CAS Number: 61956-65-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...