4-Aminobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
p-Cyanoaniline
, 4-Cyanoaniline
, p-Aminobenzonitrile
M-Clarity™ quality level = MQ100
SIAL147753-50GEA
185
GBP
SIAL147753-50G
SIAL147753-10G
4-Aminobenzonitrile, Sigma-Aldrich®
4-Aminobenzonitrile
Formula:
C₇H₆N₂ MW: 118.14 g/mol Boiling Pt: 167 °C (1.1 mmHg) Melting Pt: 84…87 °C Density: 1.274 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007821 CAS Number: 873-74-5 EINECS: 212-850-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...