3,5-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dichlorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

546933-25G 546933-5G
SIAL546933-25GEA 96.9 GBP
SIAL546933-25G SIAL546933-5G
3,5-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
3,5-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Boiling Pt: 83…84 °C (0.1 mmHg)
Melting Pt: 30…32 °C
MDL Number: MFCD00051698
CAS Number: 705-21-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR