2,4-Dichlorobenzenesulphonyl chloride

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Dichlorobenzene-1-sulphonyl chloride

D2133.10 D2133.50
XXXXD2133.10EA 0 GBP
XXXXD2133.10 XXXXD2133.50
2,4-Dichlorobenzenesulphonyl chloride
2,4-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Melting Pt: 56 °C
MDL Number: MFCD00052712
CAS Number: 16271-33-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR