4-Chlorothioanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide

002972-25G 002972-100G
FLUO002972-25GEA 0 GBP
FLUO002972-25G FLUO002972-100G
4-Chlorothioanisole
4-Chlorothioanisole
Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 44 °C (0.08 mmHg)
Melting Pt: 27…28 °C
Density: 1,2151 g/cm³ (19 °C)
MDL Number: MFCD00013643
CAS Number: 123-09-1
EINECS: 204-600-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR