2-Amino-5-chlorophenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Chloro-2-hydroxyaniline

M-Clarity™ quality level = MQ100

552224-25G 552224-5G
SIAL552224-25GEA 139 GBP
SIAL552224-25G SIAL552224-5G
2-Amino-5-chlorophenol, Sigma-Aldrich®
2-Amino-5-chlorophenol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD02093863
CAS Number: 28443-50-7
EINECS: 249-020-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR