2,3,5,6-Tetrachlorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009573-1G 009573-5G
FLUO009573-1GEA 0 GBP
FLUO009573-1G FLUO009573-5G
2,3,5,6-Tetrachlorophenyl isothiocyanate
2,3,5,6-Tetrachlorophenyl isothiocyanate
Formula: C₇HCl₄NS
MDL Number: MFCD00060365
CAS Number: 22133-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR