2-Chloro-4-fluorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-4-fluorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

638919-1G 638919-5G
SIAL638919-1GEA 44.9 GBP
SIAL638919-1G SIAL638919-5G
2-Chloro-4-fluorobenzenesulphonyl chloride, Sigma-Aldrich®
2-Chloro-4-fluorobenzenesulphonyl chloride
Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
Boiling Pt: 88…90 °C (1013 hPa)
MDL Number: MFCD00052957
CAS Number: 85958-57-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR