2-Chloro-4-fluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

013256-25G 013256-5G
FLUO013256-25GEA 0 GBP
FLUO013256-25G FLUO013256-5G
2-Chloro-4-fluoroanisole
2-Chloro-4-fluoroanisole
Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00672965
CAS Number: 2267-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR