3-Chloro-4-fluoroaniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

228583-100G 228583-25G
SIAL228583-100GEA 71 GBP
SIAL228583-100G SIAL228583-25G
3-Chloro-4-fluoroaniline, Sigma-Aldrich®
3-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 1.226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR