2-Chloro-5-fluorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009608-1G
009608-5G
FLUO009608-1GEA
0
GBP
FLUO009608-1G
FLUO009608-5G
2-Chloro-5-fluorophenyl isothiocyanate
2-Chloro-5-fluorophenyl isothiocyanate
Formula:
C₇H₃ClFNS |
CAS Number:
362690-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...