4-Chloro-2-fluorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008761-1G
008761-5G
FLUO008761-1GEA
0
GBP
FLUO008761-1G
FLUO008761-5G
4-Chloro-2-fluorophenyl isothiocyanate
4-Chloro-2-fluorophenyl isothiocyanate
Formula:
C₇H₃ClFNS |
CAS Number:
68034-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...