3-Chloro-4-fluorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-2-fluoro-5-isothiocyanatobenzene, 2-Chloro-1-fluoro-4-isothiocyanatobenzene

008759-5G 008759-25G
FLUO008759-5GEA 0 GBP
FLUO008759-5G FLUO008759-25G
3-Chloro-4-fluorophenyl isothiocyanate
3-Chloro-4-fluorophenyl isothiocyanate
Formula: C₇H₃ClFNS
MDL Number: MFCD00060700
CAS Number: 137724-66-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR