4-Chloro-3-fluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

SIAL537241-5GEA 185 GBP
SIAL537241-5G
4-Chloro-3-fluorobenzaldehyde, Sigma-Aldrich®
4-Chloro-3-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD00143288
CAS Number: 5527-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR