1,4-Dichloro-2-fluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,5-Dichlorofluorobenzene
009129-5G
009129-25G
FLUO009129-5GEA
0
GBP
FLUO009129-5G
FLUO009129-25G
1,4-Dichloro-2-fluorobenzene
1,4-Dichloro-2-fluorobenzene
Formula:
C₆H₃Cl₂F MW: 164.99 g/mol Boiling Pt: 168 °C (1013 hPa) Melting Pt: 4 °C Density: 1,383 g/cm³ (20 °C) |
MDL Number:
MFCD00060656 CAS Number: 348-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...