4-Amino-3-chlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
691445-1G
691445-5G
SIAL691445-1GEA
49.6
GBP
SIAL691445-1G
SIAL691445-5G
4-Amino-3-chlorobenzoic acid, Sigma-Aldrich®
4-Amino-3-chlorobenzoic acid
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol |
MDL Number:
MFCD00460423 CAS Number: 2486-71-7 EINECS: 219-630-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...