6-Chloroanthranilic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Amino-6-chlorobenzoic acid
M-Clarity™ quality level = MQ100
422622-10G
422622-1G
SIAL422622-10GEA
54.6
GBP
SIAL422622-10G
SIAL422622-1G
6-Chloroanthranilic acid, Sigma-Aldrich®
6-Chloroanthranilic acid
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Boiling Pt: 330 °C (1013 hPa) Melting Pt: 153…154 °C |
MDL Number:
MFCD00051530 CAS Number: 2148-56-3 EINECS: 218-416-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...