4-Amino-3,5-dichlorobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
642797-1G
642797-5G
SIAL642797-1GEA
37.2
GBP
SIAL642797-1G
SIAL642797-5G
4-Amino-3,5-dichlorobenzonitrile, Sigma-Aldrich®
4-Amino-3,5-dichlorobenzonitrile
Formula:
C₇H₄Cl₂N₂ MW: 187.03 g/mol Boiling Pt: 323 °C (1013 hPa) Melting Pt: 120 °C |
MDL Number:
MFCD00017341 CAS Number: 78473-00-4 EINECS: 278-914-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...