3,5-Dichlorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
139408-1G
139408-5G
SIAL139408-1GEA
37.2
GBP
SIAL139408-1G
SIAL139408-5G
3,5-Dichlorobenzaldehyde, Sigma-Aldrich®
3,5-Dichlorobenzaldehyde
Formula:
C₇H₄Cl₂O MW: 175.01 g/mol Boiling Pt: 215…222 °C (7 mmHg) Melting Pt: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003352 CAS Number: 10203-08-4 EINECS: 233-499-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...