3,5-Dichlorophenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

021610-5G 021610-25G
FLUO021610-5GEA 0 GBP
FLUO021610-5G FLUO021610-25G
3,5-Dichlorophenylboronic acid
3,5-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
MDL Number: MFCD00051935
CAS Number: 67492-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR