3-Cyanobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Formylbenzonitrile

M-Clarity™ quality level = MQ100

146250-1G 146250-5G
SIAL146250-1GEA 30.6 GBP
SIAL146250-1G SIAL146250-5G
3-Cyanobenzaldehyde, Sigma-Aldrich®
3-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR