2,4-Dibromobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Dibromobenzene-1-sulphonyl chloride

013065-1G 013065-5G
FLUO013065-1GEA 0 GBP
FLUO013065-1G FLUO013065-5G
2,4-Dibromobenzenesulphonyl chloride
2,4-Dibromobenzenesulphonyl chloride
Formula: C₆H₃Br₂ClO₂S
MW: 334.42 g/mol
MDL Number: MFCD03093759
CAS Number: 72256-95-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR