1-Bromo-4-nitrobenzene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Bromonitrobenzene
M-Clarity™ quality level = MQ200
167150-25G
167150-5G
SIAL167150-25GEA
112
GBP
SIAL167150-25G
SIAL167150-5G
1-Bromo-4-nitrobenzene, Sigma-Aldrich®
1-Bromo-4-nitrobenzene
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Boiling Pt: 255 °C (1013 hPa) Melting Pt: 124…126 °C Density: 1.93 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007280 CAS Number: 586-78-7 EINECS: 209-583-8 UN: 3459 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...