2,4,6-Tribromophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009577-1G 009577-5G
FLUO009577-1GEA 0 GBP
FLUO009577-1G FLUO009577-5G
2,4,6-Tribromophenyl isothiocyanate
2,4,6-Tribromophenyl isothiocyanate
Formula: C₇H₂Br₃NS
Boiling Pt: 142 °C (0.05 mmHg)
Melting Pt: 107…108 °C
MDL Number: MFCD00041039
CAS Number: 22134-11-8
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR