2-Bromo-4-fluoroanisole ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromo-4-fluoro-1-methoxybenzene

A11040.09 A11040.18
A11040.09EA 24.2 GBP
A11040.09 A11040.18
2-Bromo-4-fluoroanisole ≥98%
2-Bromo-4-fluoroanisole

Purity: 98% CAS No.: 452-08-4 Molecular Formula: C7H6BrFO Formula Weight: 205.03 Flash Point: 98°(208°F) Refractive Index: 1.5450 Boiling Point: 88-89°/1mm Beilstein Reference: 2358880 MDL No.: MFCD00012014

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 88…89 °C (1 mmHg)
Melting Pt: –27 °C
Density: 1.581
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012014
CAS Number: 452-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR