4-Bromo-3-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005219-25G 005219-100G 005219-5G
FLUO005219-25GEA 0 GBP
FLUO005219-25G FLUO005219-100G FLUO005219-5G
4-Bromo-3-fluoroaniline
4-Bromo-3-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: 72…73 °C
MDL Number: MFCD00672933
CAS Number: 656-65-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR