4-Bromo-2,6-difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005561-1G 005561-25G 005561-100G
FLUO005561-1GEA 0 GBP
FLUO005561-1G FLUO005561-25G FLUO005561-100G
4-Bromo-2,6-difluoroaniline
4-Bromo-2,6-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
MDL Number: MFCD00013389
CAS Number: 67567-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR