4-Bromo-2-fluorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009606-5G 009606-25G
FLUO009606-5GEA 0 GBP
FLUO009606-5G FLUO009606-25G
4-Bromo-2-fluorophenyl isothiocyanate
4-Bromo-2-fluorophenyl isothiocyanate
Formula: C₇H₃BrFNS
MW: 232.08 g/mol
Storage Temperature: Refrigerator
CAS Number: 81171-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR