2-Bromo-5-fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CFF839.1 005471-25G 005471-100G
CHMPCFF839.1EA 0 GBP
CHMPCFF839.1 FLUO005471-25G FLUO005471-100G
2-Bromo-5-fluorobenzoic acid
2-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00142874
CAS Number: 394-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR