4-Bromo-2-fluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

465232-25G 465232-5G
SIAL465232-25GEA 127 GBP
SIAL465232-25G SIAL465232-5G
4-Bromo-2-fluorobenzaldehyde, Sigma-Aldrich®
4-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
MDL Number: MFCD00143261
CAS Number: 57848-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR