2-Bromo-4,5-dimethoxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Bromoveratric acid

018269-1G 018269-5G
FLUO018269-1GEA 0 GBP
FLUO018269-1G FLUO018269-5G
2-Bromo-4,5-dimethoxybenzoic acid
2-Bromo-4,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.07 g/mol
MDL Number: MFCD00017542
CAS Number: 6286-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR