1-Methylisatin, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Methylindoline-2,3-dione, 1-Methyl-2,3-dihydroindole-2,3-dione, N-Methylindoline-2,3-dione, N-Methylisatin, 1-Methyl-1H-indole-2,3-dione, N-Methyl-1H-indole-2,3-dione, N-Methyl-2,3-dihydroindole-2,3-dione
183075-10G
183075-1G
SIAL183075-10GEA
85
GBP
SIAL183075-10G
SIAL183075-1G
1-Methylisatin, Sigma-Aldrich®
1-Methylisatin
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Melting Pt: 139 °C Density: 1.4 g/cm³ (20 °C) |
MDL Number:
MFCD00005812 CAS Number: 2058-74-4 EINECS: 218-164-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...