4-Methylmorpholine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: NMM, N-Methylmorpholine

M-Clarity™ quality level = MQ200

67870-1L 67870-250ML
SIAL67870-1LEA 140 GBP
SIAL67870-1L SIAL67870-250ML
4-Methylmorpholine, Sigma-Aldrich®
4-Methylmorpholine
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.922 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006175
CAS Number: 109-02-4
EINECS: 203-640-0
UN: 2535
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR