Anthracene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

SIAL694959-5GEA 48.9 GBP
SIAL694959-5G SIAL694959-25G
Anthracene, Sigma-Aldrich®
Anthracene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 111…112 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001240
CAS Number: 120-12-7
EINECS: 204-371-1
Merck Index: 13,00687
https://uk.vwr.com/stibo/web/std.lang.all/31/13/743113.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR