2,5-Dimethoxycinnamic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(2,5-Dimethoxyphenyl)acrylic acid, 2',5'-Dimethoxycinnamic acid, trans-2,5-Dimethoxycinnamic acid

M-Clarity™ quality level = MQ100

143596-100G 143596-25G
SIAL143596-100GEA 232 GBP
SIAL143596-100G SIAL143596-25G
2,5-Dimethoxycinnamic acid, Sigma-Aldrich®
2,5-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 150 °C
MDL Number: MFCD00004378
CAS Number: 10538-51-9
EINECS: 234-114-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR