1-Tetradecene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIALT9805-1LEA 63.9 GBP
SIALT9805-1L
1-Tetradecene, Sigma-Aldrich®
1-Tetradecene
Formula: C₁₄H₂₈
MW: 196.38 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: –13 °C
Density: 0.7708 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008981
CAS Number: 1120-36-1
EINECS: 214-306-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR