1-Hexadecene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SIALH2131-100MLEA 199 GBP
SIALH2131-100ML
1-Hexadecene, Sigma-Aldrich®
1-Hexadecene
Formula: C₁₆H₃₂
MW: 224.43 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 4 °C
Density: 0,7811 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00008991
CAS Number: 629-73-2
EINECS: 211-105-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR