trans-2-Nonenal, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(E)-2-Nonenal
M-Clarity™ quality level = MQ100
255653-25G
255653-5G
SIAL255653-25GEA
299
GBP
SIAL255653-25G
SIAL255653-5G
trans-2-Nonenal, Sigma-Aldrich®
trans-2-Nonenal
Formula:
C₉H₁₆O MW: 140.23 g/mol Boiling Pt: 100 °C (1013 hPa) Density: 0.8505 g/cm³ (15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007012 CAS Number: 18829-56-6 EINECS: 242-609-6 Merck Index: 12,06771 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...