trans-1,4-Dibromo-2-butene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene

M-Clarity™ quality level = MQ100

D39207-100G D39207-25G
SIALD39207-100GEA 333 GBP
SIALD39207-100G SIALD39207-25G
trans-1,4-Dibromo-2-butene, Sigma-Aldrich®
trans-1,4-Dibromo-2-butene
Formula: C₄H₆Br₂
MW: 213.9 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 50…53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000249
CAS Number: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR