11-Bromo-1-undecene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001789-5G 001789-25G
FLUO001789-5GEA 0 GBP
FLUO001789-5G FLUO001789-25G
11-Bromo-1-undecene
11-Bromo-1-undecene
Formula: C₁₁H₂₁Br
MW: 233.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040825
CAS Number: 7766-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR